missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Acura electro™ 956 12-Channel Pipetters - Basic Package

Numéro de catalogue. p-4450887
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 200 μL
2.5 to 50 μL
20 to 350 μL
Accuracy:
±0.9 to <±0.6%
±1 to <±0.6%
±1 to <±0.8%
±3.5 to <±1%
Increments:
0.05μL
0.25μL
1μL
5μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric) Accuracy Increments
03410017 2.5 to 50 μL ±1 to <±0.8% 0.25μL
03410016 0.5 to 10 μL ±3.5 to <±1% 0.05μL
03410019 20 to 350 μL ±1 to <±0.6% 5μL
03410018 10 to 200 μL ±0.9 to <±0.6% 1μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 03410017 Fournisseur DWK Life Sciences Code fournisseur W870934A
Il en reste null

Comprend

Acura electro basic package includes electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, and pipette tip samples.

Large selection of 12-channel pipettes

360° rotation of multichannel volumetric module for best working position

  • Microprocessor-controlled motor
  • Rechargeable battery pack
  • Also fits any single channel volumetric module
  • Models cover the volume range 0.5 to 350μL

Spécifications

Accuracy ±1 to <±0.8%
Battery Type Rechargeable
Compatible Tips Universal fit pipette tip
Model 956
Number of Channels 12
Product Line Acura Electro
Type Pipetters Basic Package
Volume (Metric) 2.5 to 50 μL
Display Digital
Finger Style Ergonomic
Includes Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack and pipette tip samples
Increments 0.25μL
Warranty 2 Years
Afficher plus de résultats Afficher moins de résultats