missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wheat Germ Agglutinin (WGA)

Numéro de catalogue. W11263
Encompass
Modifier l'affichage
Click to view available options
Conjugué:
Alexa Fluor 350
Alexa Fluor 350, Oregon Green 488, Tetramethylrhodamine, Texas Red
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 594
Alexa Fluor 633
Alexa Fluor 647
Alexa Fluor 680
Alexa Fluor Plus 405
Alexa Fluor Plus 568
Alexa Fluor Plus 770
Fluorescein
Oregon Green 488
Qdot 655
Tetramethylrhodamine
Texas Red-X
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Conjugué
W11263 Alexa Fluor 350
W7024 Alexa Fluor 350, Oregon Green 488, Tetramethylrhodamine, Texas Red
W11261 Alexa Fluor 488
W32464 Alexa Fluor 555
W11262 Alexa Fluor 594
W21404 Alexa Fluor 633
W32466 Alexa Fluor 647
W32465 Alexa Fluor 680
W56132 Alexa Fluor Plus 405
W56133 Alexa Fluor Plus 568
W56134 Alexa Fluor Plus 770
W834 Fluorescein
W6748 Oregon Green 488
Q12021MP Qdot 655
W849 Tetramethylrhodamine
W21405 Texas Red-X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Numéro de catalogue. W11263 Fournisseur Invitrogen™ Code fournisseur W11263

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) and Alexa Fluor, Alexa Fluor Plus, and other dyes. Fluorescent WGA conjugates bind to carbohydrates and are used for various cell biology applications such as plasma membrane labeling and cell painting assays.

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates. These lectins can bind to carbohydrates and are available conjugated to Alexa Fluor™, Alexa Fluor™ Plus, and other fluorescent dyes. Fluorescent wheat germ agglutinin conjugates are valuable tools in molecular and cell biology research, enabling researchers to label the plasma membrane in fluorescence imaging and cell painting assays, and study and analyze glycosylation patterns and glycan-mediated processes in cells and tissues.

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) with Alexa Fluor, Alexa Fluor Plus, and other dyes. WGA is a cell impermeant stain that selectively binds to N‐acetylglucosamine and N‐acetylneuraminic acid (sialic acid) residues, which are often found on cell membranes. Fluorescent WGA conjugates provide selective labeling of the plasma membrane with minimal background in many cell types that is retained after formaldehyde fixation and permeabilization with Triton X-100.

These fluorescent lectin conjugates can also be used to label fixed cells; however, to avoid labeling intracellular components, formaldehyde-fixed cells should not be permeabilized before labeling. Fluorescent WGA conjugates are used as plasma membrane stains along with other cellular markers in cell painting assays to provide a phenotypic readout of cell health or cytotoxicity. The Wheat Germ Agglutinin, Alexa Fluor 555 Conjugate is included in the Image-iT Cell Painting Kit (Cat. Nos. I65000 and I65500).

WGA conjugates are also used as retrograde tracers for neuronal tracing experiments and have been shown to cross synapses. These fluorescent lectins are applicable in microbiology studies to label yeast bud scars, the cell membrane of gram-positive but not gram-negative bacteria, and chitin in fungal cell walls. In solution, WGA exists as a heterodimer with a molecular weight of approximately 38,000 Daltons and is normally cationic under physiological conditions. Our WGA conjugates have been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), flow cytometry (FC), and a wide range of chemical, biochemical and immunological assays.

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates with options covering the entire wavelength range. The Wheat Germ Agglutinin Sampler Kit (Cat. No. W7024) includes introductory samples of four fluorescent WGAs: Alexa Fluor 350, Oregon Green 488, tetramethylrhodamine, and Texas Red-X conjugates. The red-fluorescent Alexa Fluor 594 wheat germ agglutinin (WGA) conjugate is also included in the Image-iT LIVE Plasma Membrane and Nuclear Labeling Kit (Cat. No. I34406). The Texas Red-X WGA conjugate can be purchased in the ViaGram Red+ Bacterial Gram Stain and Viability Kit (Cat. No. V7023) to differentiate gram-positive and gram-negative bacteria.

Wheat germ agglutinin (WGA) conjugates fluorescence excitation/emission
W11261 WGA, Alexa Fluor 488 conjugate: 495 nm/519 nm
W11263 WGA, Alexa Fluor 350 conjugate: 346 nm/442 nm
W32464 WGA, Alexa Fluor 555 conjugate: 555 nm/580 nm
W11262 WGA, Alexa Fluor 594 conjugate: 590 nm/617 nm
W21404 WGA, Alexa Fluor 633 conjugate: 632 nm/647 nm
W32466 WGA, Alexa Fluor 647 conjugate: 650 nm/665 nm
W32465 WGA, Alexa Fluor 680 conjugate: 679 nm/702 nm
W56132 WGA, Alexa Fluor Plus 405 conjugate: 408 nm/450 nm
W56133 WGA, Alexa Fluor Plus 568 conjugate: 562 nm/583 nm
W56134 WGA, Alexa Fluor Plus 770 conjugate: 770 nm/797 nm
W834 WGA, fluorescein conjugate: 494 nm/518 nm
W6748 WGA, Oregon Green 488 conjugate: 496 nm/524 nm
W849 WGA, tetramethylrhodamine conjugate: 555 nm/580 nm
W21405 WGA, Texas Red-X conjugate: 595 nm/615 nm

Spécifications

À utiliser avec (application) Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Cell Painting
Quantité 5 mg
Sources Wheat germ
Nom usuel Wheat Germ Agglutinin, Alexa Fluor™ 350 Conjugate
Conjugué Alexa Fluor 350
Espèces Wheat
Recombinant Native
Étiquette de la protéine None
Contenu et stockage Store in freezer (-5 to -30°C) and protect from light.
Shipping condition (Condition d'expédition) Room Temperature
Système d’expression Wheat germ
Famille de protéines Lectins
Forme de protéine Heterodimer
Forme Lyophilized
Sous-type de protéine Agglutinin
Gamme de produit Alexa Fluor
Grade de qualité ou de pureté See Certificate of Analysis
Protéine Fluorescent lectins
Type de ligand N-acetylglucosamine and N-acetylneuraminic acid (sialic acid) residues
Afficher plus de résultats Afficher moins de résultats

For Research Use Only. Not for use in diagnostic procedures.