Learn More
Cytiva Whatman™ Polyester Drain Discs
Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.
Supplier: Cytiva 231100
Description
Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.
For use with membrane hardware where extra support is needed for improved flow rate and throughput. Can be used as a separator between membranes in serial stack filtration applications.
- Chemically inert polyester
- 100μm Thickness
- Binder-free
- Available in several diameters
Specifications
| Polyester | |
| 100 μm | |
| 47 mm |