missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Phenolphthalein Test Paper on Reel
Description
Combines ease-of-use with accuracy and consistency for determination of the neutralization point in weak acid/strong alkali reactions.
The convenience of using indicator papers for the rapid determination of pH values has led to many applications in laboratories and industry
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Convenient and portable for field use
- Economical
Specifications
Specifications
| Color Indicator | Phenolphthalein |
| pH Range | 8.3 to 10 |
| For Use With (Application) | For determination of the neutralization point in weak acid/strong alkali reactions |
| Quantity | Each |
| Type | Phenolphthalein Test Paper |