Learn More
Cytiva Whatman™ Lead Acetate Test Paper Dispenser
Description
Dispenser for rapid qualitative test paper to detect hydrogen sulfide
Lead Acetate Test Paper Detections
- As little as 5ppm of atmospheric or gas stream H2S
- As little as 4ppm H202 by preblackening paper with H2S
Spécifications
Spécifications
| For Use With (Application) | For rapid qualitative test paper to detect hydrogen sulfide, detect ∼5ppm of H2S in atmosphere or gas stream |
| Quantity | Each |
| Type | Lead Acetate Test Paper Dispenser |