missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN QIAcard FTA Elute Indicating Micro
For collection, stabilization, processing, transport and archiving of biological samples. 1 sample area per card.
$440.18 - $1,841.52
Specifications
| Content And Storage | Room Temperature |
|---|---|
| Description | Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix. |
| For Use With (Application) | Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection |
| Includes | Inert dye |
| Product Type | Indicating FTA Elute Micro Card |
Description
- FTA Elute technology for convenient water based elution of nucleic acids.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
| Room Temperature | |
| Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection | |
| Indicating FTA Elute Micro Card | |
| 15 to 30 min |
| Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix. | |
| Inert dye | |
| 40 μL |