Learn More
QIAGEN QIAcard FTA Elute Indicating Micro
For collection, stabilization, processing, transport and archiving of biological samples. 1 sample area per card.
Supplier:  QIAGEN WB120412
 View more versions of this product
Catalog No. 09800996
Only null left
Description
Description
  • FTA Elute technology for convenient water based elution of nucleic acids.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Chemical Identifiers
Room Temperature
Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection
Indicating FTA Elute Micro Card
15 to 30 min
Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix.
Inert dye
40 μL
Specifications
Room Temperature
Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection
Indicating FTA Elute Micro Card
40 μL
Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix.
Inert dye
25 Pk.
15 to 30 min
Product Suggestions
Product Suggestions