Learn More
QIAGEN Foam Swabs, Sterile
Catalog No. 09923375
Change view
Click to view available options
Quantity:
100 Pk.
Catalog No. Quantity
09923375 100 Pk.
1 options
Catalog No. 09923375 Supplier QIAGEN Supplier No. WB100032
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For buccal and saliva samples.
  • Sterile foam swabs for easy collection of buccal cells and saliva and transfer to e.g. QIAcard FTA card formats.
  • The nonabrasive head of the foam tipped applicator is the same size as the sample area on QIAcard FTA Cards to facilitate sample application.
Specifications
Type Foam-tipped Applicator
For Use With (Application) For the collection of saliva and buccal cells
Quantity 100 Pk.