Research Products Corporation WetGrip Micro-Tube Laser Printer Labels, Fits 1.5ml Tubes, 1.28 x 0.5 Inches, 1700 per Package
Catalog No. 50998652
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.
Catalog No.
50998652
Supplier
Research Products CorporationSupplier No.
RPI 247175
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.