Research Products Corporation WetGrip Micro-Tube Laser Printer Labels, Fits 1.5 -2.0ml Tubes, 1.50 x 0.75 Inches, 1200 per Package
Numéro de catalogue. 50998654
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.
Numéro de catalogue.
50998654
Fournisseur
Research Products CorporationCode fournisseur
RPI 247177
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.