Learn More
Mtc Bio Western Blot Boxes, 6/PK
Numéro de catalogue. 501870987
Encompass
This item has a minimum qty of 2 per supplier requirements.
MultiBox™, 4 compartments, 85 x 85 x 30mm each (11/2 x 2 13/16 x 11/8 in.), for various gels including half-mini protein gels
Numéro de catalogue. 501870987 Fournisseur Mtc Bio Code fournisseur B1204*
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This item has a minimum qty of 2 per supplier requirements.
MultiBox™, 4 compartments, 85 x 85 x 30mm each (11/2 x 2 13/16 x 11/8 in.), for various gels including half-mini protein gels