missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Polyurethane Palm

Fit and comfortable protection

$111.49 - $111.49

Specifications

Cut Resistance Rating ANSI A3
Product Type Cut-resistant Gloves
For Use With (Application) Protection from lacerations and superior resistance against cuts and for grip and abrasion resistance
Glove Cuff Style Knit
View More Specs

Products 6
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
19148740
View Documents
Wells Lamont™
Y9284XS
Yellow X-Small
Dozen of 12 for $111.49
Case of 12 Dozen for $1,146.84
Only null left
 
19148797
View Documents
Wells Lamont™
Y9284S
Red Small
Dozen of 12 for $111.49
Case of 12 Dozen for $1,146.84
Only null left
 
19148796
View Documents
Wells Lamont™
Y9284M
Green Medium
Dozen of 12 for $111.49
Case of 12 Dozen for $1,146.84
Only null left
 
19148798
View Documents
Wells Lamont™
Y9284L
Brown Large
Dozen of 12 for $111.49
Case of 12 Dozen for $1,146.84
Only null left
 
19148831
View Documents
Wells Lamont™
Y9284XL
Blue X-Large
Dozen of 12 for $111.49
Case of 12 Dozen for $1,146.84
Only null left
 
19148832
View Documents
Wells Lamont™
Y9284XXL
Black 2X-Large
Dozen of 12 for $111.49
Case of 12 Dozen for $1,146.84
Only null left
 
Description

Description

  • Features highly cut-resistant fiber that protects against lacerations
  • Polyurethane palm coating offers grip and abrasion resistance
  • Dark-colored shell conceals dirt and grime
  • Knit wrist construction prevents dirt and debris from entering the glove
  • Color: Gray

Specifications

Specifications

ANSI A3
Cut-resistant Gloves
Protection from lacerations and superior resistance against cuts and for grip and abrasion resistance
Knit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'