Learn More
Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Polyurethane Palm
Fit and comfortable protection
Supplier:  Wells Lamont™ Y9284XL
 View more versions of this product
Catalog No. 19148831
Only null left
Explore available promotions
Description
Description
  • Features highly cut-resistant fiber that protects against lacerations
  • Polyurethane palm coating offers grip and abrasion resistance
  • Dark-colored shell conceals dirt and grime
  • Knit wrist construction prevents dirt and debris from entering the glove
  • Color: Gray
Specifications
Specifications
Gray
Polyurethane
X-Large
Blue
ANSI A3
Cut-resistant Gloves
Protection from lacerations and superior resistance against cuts and for grip and abrasion resistance
Knit
Product Suggestions
Product Suggestions