missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Polyurethane Palm

Catalog No. 19148740
Change view
Click to view available options
Glove Cuff Color:
Black
Blue
Brown
Green
Red
Yellow
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Color Size
19148740 Yellow X-Small
19148797 Red Small
19148796 Green Medium
19148798 Brown Large
19148831 Blue X-Large
19148832 Black 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19148740 Supplier Wells Lamont™ Supplier No. Y9284XS
Only null left

Fit and comfortable protection

  • Features highly cut-resistant fiber that protects against lacerations
  • Polyurethane palm coating offers grip and abrasion resistance
  • Dark-colored shell conceals dirt and grime
  • Knit wrist construction prevents dirt and debris from entering the glove
  • Color: Gray

Specifications

Coating Material Polyurethane
Cut Resistance Rating ANSI A3
Material Knit
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Protection from lacerations and superior resistance against cuts and for grip and abrasion resistance
Glove Cuff Color Yellow
Glove Cuff Style Knit
Hem Color Yellow
Series GuardTech™ 3