Learn More
Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Polyurethane Palm
Fit and comfortable protection
Supplier: Wells Lamont™ Y9284L
Description
- Features highly cut-resistant fiber that protects against lacerations
- Polyurethane palm coating offers grip and abrasion resistance
- Dark-colored shell conceals dirt and grime
- Knit wrist construction prevents dirt and debris from entering the glove
- Color: Gray
Specifications
| Gray | |
| Polyurethane | |
| Large | |
| Brown |
| ANSI A3 | |
| Cut-resistant Gloves | |
| Protection from lacerations and superior resistance against cuts and for grip and abrasion resistance | |
| Knit |