missing translation for 'onlineSavingsMsg'
Learn More

Wax Paraffin, Beads, Spectrum™ Chemical

Catalog No. 18614673
Change view
Click to view available options
Quantity:
1 lb.
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614673 1 lb. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18614673 Supplier Spectrum Chemical Mfg Cor Supplier No. W10181LB
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Versatile paraffin wax in bead form, suitable for industrial applications including electrical insulation, lubrication, and candle making.

Chemical Identifiers

CAS 8002-74-2
Molecular Formula C21H27NO3
Molecular Weight (g/mol) 341.45
InChI Key JWHAUXFOSRPERK-UHFFFAOYNA-N
IUPAC Name 1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
SMILES CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1

Specifications

Acidity 1
CAS Max % 1
Melting Point 130°F to 135°F
Packaging Poly Bottle
Quantity 1 lb.
Saponification Value 1
Grade Ungraded
Physical Form Beads
Chemical Name or Material Wax Paraffin, Beads

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.