missing translation for 'onlineSavingsMsg'
Learn More

Water, USP Purified, Nonsterile, Ricca Chemical

Catalog No. 361015694
Change view
Click to view available options
Quantity:
55 gal.
Packaging:
Poly Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361015694 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361015694 Supplier Ricca Chemical Company Supplier No. R919100055E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Percent Purity 100%
Grade USP Purified
Packaging Poly Drum
Quantity 55 gal.
Physical Form Liquid
Chemical Name or Material Water, Non-Sterile
Show More Show Less