missing translation for 'onlineSavingsMsg'
Learn More

Water, USP/EP Purified, Ricca Chemical

Catalog No. 91905
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Hard Pack
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
91905 20 L Cubitainer
91901G 4 L Amber Glass Bottle
91901 4 L Natural Poly Bottle
919025 10 L Cubitainer
91905HP 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 91905 Supplier Ricca Chemical Company Supplier No. 91905

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water, USP/EP Purified

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

pH 5.0 to 7.0
Color Clear, Colorless
CAS Min % 98.0
CAS Max % 100.0
Conductivity max. 1 μS/cm
Endotoxin max. 0.25 EU/mL at time of manufacture
Synonym dihydrogen oxide, dihydrogen monoxide
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Percent Purity 100%
Grade EP/USP
Packaging Cubitainer
Residue after Evaporation max. 0.001 %
Sterility Non-sterile
Quantity 20 L
Chemical Name or Material Water
Show More Show Less
ShelfLife 12 months