missing translation for 'onlineSavingsMsg'
Learn More

Water, Reagent Grade; OmniSolv™, MilliporeSigma™

Catalog No. 361042007
Change view
Click to view available options
Quantity:
20 L
Packaging:
Plastic Barrel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361042007 20 L Plastic Barrel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361042007 Supplier Technidata Supplier No. WX0004NP20

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Used for testing USP/NF monographs
  • For HPLC Gradient Analysis, Spectrophotometry, Gas Chromatography

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH Neutral
Melting Point 0°C
Boiling Point 100°C (1013 hPa)
Formula Weight 18.02g/mol
Packaging Plastic Barrel
Quantity 20 L
Physical Form Liquid