missing translation for 'onlineSavingsMsg'
Learn More

Water, MilliporeSigma

Catalog No. 1037281002
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1037281002 1 L
1037282002 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1037281002 Supplier Technidata Supplier No. 1037281002

LiChrosolv™ solvents exhibit high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them ideal for reproducible separations.

Chemical Identifiers

CAS 7732-18-5
Molecular Weight (g/mol) 18.02 g/mol

Specifications

Grade UHPLC-MS
Packaging Borosilicate Glass Bottle
Quantity 1 L
Chemical Name or Material Water