missing translation for 'onlineSavingsMsg'
Learn More

Water for Irrigation, USP, Sterile Grade, Cytiva Hyclone™

Catalog No. WIRUSP5XM
Change view
Click to view available options
Quantity:
6 x 500 mL
4 x 4 L
6 x 1 L
Packaging:
HDPE Jug
PET Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
WIRUSP5XM 6 x 500 mL PET Plastic Bottle
WIRUSP1X6 6 x 1 L PET Plastic Bottle
Z0304 4 x 4 L HDPE Jug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. WIRUSP5XM Supplier Cytiva Supplier No. WIRUSP5XM
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Water for Irrigation, USP is a sterile, hypotonic, non-pyrogenic fluid or solvent. It is prepared by terminal distillation and contains no antimicrobial or bacteriostatic agents or added buffers.

  • The pH is 5.0 to 7.0
  • Complies with the USP monograph for water for irrigation
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications
  • Widely used in applications where a high quality sterile water is required

For Manufacturing or Research Use only, not intended for Diagnostic or Therapeutic Use.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Endotoxin <0.25 EU/mL
Grade USP
Packaging PET Plastic Bottle
Sterility Sterile
Certificate of Analysis Yes
Quantity 6 x 500 mL
ShelfLife 36 Months
Recommended Storage 2-30°C