missing translation for 'onlineSavingsMsg'
Learn More

Water, DIUF, Fisher Chemical™

Catalog No. W24
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
W24 4 L Poly Bottle
W220 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. W24 Supplier Thermo Fisher Scientific Supplier No. W2 4
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water, DIUF

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Packaging Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Density 1.000g/cm³
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water, D.I.U.F.
Show More Show Less