missing translation for 'onlineSavingsMsg'
Learn More

Water, Distilled, Reagent Grade, Ricca Chemical

Catalog No. R918000055E
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Cubitainer
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R918000055E 55 gal. Poly Drum
918032 1 L Amber Glass Bottle
91801 4 L Amber Glass Bottle
91805 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R918000055E Supplier Ricca Chemical Company Supplier No. R918000055E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Color Colorless
Conductivity max. 1.5 μS/cm
Synonym dihydrogen oxide
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 100%
Grade Reagent
Packaging Poly Drum
Sterility Non-sterile
Quantity 55 gal.
Chemical Name or Material Water, Distilled
Show More Show Less
ShelfLife 18 Months