missing translation for 'onlineSavingsMsg'
Learn More

Water, for HPLC and Spectrophotometry, Solstice

Catalog No. 6002652
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6002652 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002652 Supplier Solstice Supplier No. 3654

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm, 0.01 AU max. at 190 nm, 0.01 AU max. at 200 nm, 0.005 AU max. at 250 nm
Boiling Point 100°C
CAS Max % 1
Beilstein 2050024
Formula Weight 18
Grade HPLC
Packaging Glass Bottle
Density 0.9971 g/cm3
Quantity 4 L
Chemical Name or Material Water, B&J Brand, for HPLC and Spectrophotometry
ShelfLife 547 days from date of manufacture