missing translation for 'onlineSavingsMsg'
Learn More

Water, ACS reagent

Catalog No. AC434400050
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434400050 5 L Plastic Bottle
AC434400010 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434400050 Supplier Thermo Scientific Chemicals Supplier No. 434400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7732-18-5 | H2O | 18.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Absorbance Passes Test, 0.01 max. at 200nm, 0.005 max. at 300nm, 0.01 max. at 210nm, 0.005 max. at 254nm
Color White
Boiling Point 100°C
Conductivity 2μS/cm Max.
Linear Formula H2O
Merck Index 15,10237
Formula Weight 18.02
Grade ACS Reagent
Packaging Plastic Bottle
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 5 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Crystalline Powder
Chemical Name or Material Water, For analysis ACS
Show More Show Less
Hazard Category
EINECSNumber 231-791-2

RUO – Research Use Only