missing translation for 'onlineSavingsMsg'
Learn More

Water, ACS Grade, Vion™

Numéro de catalogue. p-200135625
Encompass
Change view
Click to view available options
Quantity:
60 mL
125 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
11178109 125 mL
11178110 1 L
11178111 250 mL
11178112 500 mL
11178113 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 11178109 Fournisseur Aldon Corporation Code fournisseur VNWW7000125ML
Il en reste null

Vion ACS Reagent grade water is guaranteed to meet the purity standards set by the American Chemical Society (ACS). It is a high-quality, highly purified water with minimal contaminants, and is ideal for use in laboratory settings for sensitive chemical analyses where extreme purity is required.

Specifications:

  • Conductivity ≤ 2.0 μS/cm
  • Silicate (as SiO2) ≤ 0.01 ppm
  • Heavy Metals (as Pb) ≤ 0.01 ppm
  • Substances Reducing Permanganate - Passes Test
  • Chloride (Cl) ≤ 0.4 ppm
  • Nitrate (NO3) ≤ 0.4 ppm
  • Phosphate (PO4) ≤ 1 ppm
  • Sulfate (SO4) ≤ 1 ppm
  • UV A300 nm ≤ 0.005
  • UV A254 nm ≤ 0.005
  • UV A210 nm ≤ 0.010
  • UV A200 nm ≤ 0.010
  • LC Absorbance - Passes Test
  • LC Gradient Elution - Passes Test

Identifiants chimiques

Molecular Weight (g/mol) 18.02 g/mol

Spécifications

CAS 7732-18-5
Percent Purity 100%
Grade ACS
Quantity 125 mL
Chemical Name or Material Water