missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Waste Disposal Labels - Plastic

Catalog No. 19111186
Change view
Click to view available options
Dimensions (L x W):
6 x 6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19111186 6 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19111186 Supplier Brady™ Supplier No. BRD63240

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use these durable labels to easily identify and sort waste for disposal.

Made of plastic

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Description Label
Adhesive Type Self-adhesive
Color Black on White
Format Label
Material Plastic
Printable Sides Single-side Printable
Product Type Label
Languages English
Quantity 500/Roll
Shape Rectangle
Size (English) 6 x 6 in.
Dimensions (L x W) 6 x 6 in.
Width (English) 6 in.
Width (Metric) 157.48 mm
Height (English) 6 in.
Show More Show Less