missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Warning Labels, Pictogram: Flammable Material
Feature pictograms that indicate hazards for flammable material. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.
$96.66 - $203.57
Specifications
| Description | Pictogram: Flammable Material |
|---|---|
| Adhesive Type | Acrylic, Permanent, Cold-temperature |
| Color | Black, Yellow |
| Material | Pressure-sensitive Vinyl (B-946) |
| For Use With (Application) | Indoor/Outdoor |
| Catalog Number | Mfr. No. | Size (English) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size (English) | Price | Quantity & Availability | |||||
|
19107252
|
Brady™
60181 |
0.5 x 0.5 in. |
Pack of 10 Each for $96.66
|
|
|||||
|
19107259
|
Brady™
60192 |
1 x 1 in. |
Pack of 10 Each for $103.17
|
|
|||||
|
19107266
|
Brady™
60203 |
2 x 2 in. |
Pack of 10 Each for $180.33
|
|
|||||
|
19107273
|
Brady™
60214 |
4 x 4 in. |
Pack of 10 Each for $120.83
|
|
|||||
|
19107280
|
Brady™
60225 |
8 x 8 in. |
Pack of 10 Each for $203.57
|
|
|||||
Description
- Made with highly visible B-946 bright colored vinyl
- Adheres to any clean, dry surface
- Available in 5 different sizes
Specifications
| Pictogram: Flammable Material | |
| Black, Yellow | |
| Indoor/Outdoor | |
| 0.004 in. |
| Acrylic, Permanent, Cold-temperature | |
| Pressure-sensitive Vinyl (B-946) | |
| Triangle |