missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Warning Labels, Pictogram: Flammable Material

Feature pictograms that indicate hazards for flammable material. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.

Supplier:  Brady™ 60214

 View more versions of this product

Catalog No. 19107273


Only null left

Description

Description

  • Made with highly visible B-946 bright colored vinyl
  • Adheres to any clean, dry surface
  • Available in 5 different sizes
Specifications

Chemical Identifiers

Pictogram: Flammable Material
Black, Yellow
Indoor/Outdoor
0.004 in.
Acrylic, Permanent, Cold-temperature
Pressure-sensitive Vinyl (B-946)
Triangle

Specifications

Pictogram: Flammable Material
Black, Yellow
Warning Label
Triangle
0.004 in.
Acrylic, Permanent, Cold-temperature
Pressure-sensitive Vinyl (B-946)
Indoor/Outdoor
4 x 4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions