missing translation for 'onlineSavingsMsg'
Learn More

Conair™ Waring™ Laboratory Blenders: Single Speed

Catalog No. 1450950
Change view
Click to view available options
Container Material:
Glass
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Container Material
1450950 Stainless Steel
1450910 Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1450950 Supplier Conair™ Supplier No. 700S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Base, container, 3-wire grounded cord and plug

Meet the requirements of the most demanding mixing and blending applications

  • Single-speed
  • Cloverleaf design
  • Choice of glass and/or stainless-steel containers
  • Sturdy, diecast metal bases and stainless-steel blades
  • All durably constructed for years of dependable service
  • Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
  • Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
  • Two-piece vinyl and styrene lids

Order Info

Electrical requirements for all models: 115/120V 50/60Hz

Certifications

UL listed (Select models only; see ordering chart)

Specifications

Amperage 3.6 A
Speed 22,000 rpm
Voltage 115/120 V
Electrical Requirements 115/120 V, 50/60 Hz
Horsepower 0.5 hp
Wattage 350 W
Capacity (Metric) 1000 mL
Sample Volume (English) 34 oz.
Depth (English) 7 in.
Depth (Metric) 17.78 cm
Height (English) 16 in.
Height (Metric) 41 cm
Width (English) 8 in.
Width (Metric) 20.32 cm
Base Material Epoxy-coated Metal
Container Material Stainless Steel
Includes Base, container, 3-wire grounded cord and plug
Packaging Quantity 1 Ea.
Frequency 50/60 Hz
Show More Show Less