missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Vytac™ Formalin Disposal System, Vytac

Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Epredia™ Vytac™ Formalin Disposal System.

Supplier:  Epredia™ 5401

 Afficher plus de versions de ce produit

Numéro de catalogue. 22046332



Il en reste null

Description

Description

Disposal Solution:
  • Breaks down formalin into inert components
  • Neutralizes four parts formalin waste per one part Vytac
  • When used as directed, neutralizes more than 99% of the waste formalin collected
Note: Vytac GNF will neutralize gluteraldehyde
Spécifications

Spécifications

Vytac
Vytac; 1 Gal. (3.79L)
1 Gal. (3.79 L)
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions