missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0357017 hydrochloride

Catalog No. 429510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
429510 10 mg
429550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 429510 Supplier Tocris Bioscience™ Supplier No. 4295/10

Positive allosteric modulator of M1 receptors

Positive allosteric modulator of muscarinic M1 receptors (EC50 = 198 nM). Displays no activity at M2-M5 at concentrations up to 30 μM. Potentiates NMDA receptor currents in hippocampal neurons; activity reverses cognitive decifits in a rodent model of hippocampal-dependent memory. CNS penetrant.

Chemical Identifiers

CAS 1135242-13-5
Molecular Formula C18H28ClN3O3

Specifications

Quantity 10 mg
Formula Weight 369.89
Purity >98%
Molecular Weight (g/mol) 369.89
Chemical Name or Material VU 0357017 hydrochloride
Recommended Storage Store at +4°C