CAS RN 1135242-13-5
IUPAC Name:
ethyl 4-({2-[(2-methylphenyl)formamido]ethyl}amino)piperidine-1-carboxylate hydrochloride
Synonyms:
ethyl 4-({2-[(2-methylphenyl)formamido]ethyl}amino)piperidine-1-carboxylate hydrochloride
Molecular Weight (g/mol):
369.89
Molecular Formula:
C18H28ClN3O3
InChi Key:
XKJQVUIXSBOCPP-UHFFFAOYSA-N
SMILES:
Cl.CCOC(=O)N1CCC(CC1)NCCNC(=O)C1=CC=CC=C1C
1
–
1
of
1
results
1
Tocris Bioscience™ VU 0357017 hydrochloride
CAS: 1135242-13-5 Molecular Formula: C18H28ClN3O3