missing translation for 'onlineSavingsMsg'
Learn More

VSV-Glycoprotein Mouse anti-Virus, Clone: 8G5F11, MilliporeSigma™

Numéro de catalogue. MABF2337100
Modifier l'affichage
Click to view available options
Quantité:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MABF2337100 100 μg
MABF233725 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MABF2337100 Fournisseur MilliporeSigma Code fournisseur MABF2337100UG

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

VSV-Glycoprotein Monoclonal antibody specifically detects VSV-Glycoprotein in Virus samples. It is validated for Flow Cytometry, Inhibition Assay, Neutralization, Radioimmune Assays, Western Blotting.

Spécifications

Antigène VSV-Glycoprotein
Applications Flow Cytometry, Inhibition Assays, Neutralization, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 8G5F11
Conjugué Unconjugated
Formule Purified mouse monoclonal antibody IgG2a in PBS without azide.
Espèces hôtes Mouse
Immunogène Vesicular stomatitis virus, Indiana serotype
Méthode de purification Protein G purified
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Virus
Contenu et stockage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Forme Purified
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats