missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Vortexer Mixer, Blue

Numéro de catalogue. 14127521
Change view
Click to view available options
Plug Type:
CE
JP Plug
US Plug
Electrical Requirements:
110/120 V, 50/60 Hz
220/240 V, 50/60 Hz
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Plug Type Electrical Requirements
14127521 CE 220/240 V, 50/60 Hz
14127520 US Plug 110/120 V, 50/60 Hz
14127524 JP Plug 220/240 V, 50/60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 14127521 Fournisseur Heathrow Scientific™ Code fournisseur HS120212
Il en reste null

The Vortexer is a multi-purpose, and maximum convenience unit. This sleek and modern designed Vortexer Mixer is capable of meeting the mixing requirements of the most popular test tubes used in today’s lab without the need to swap out mixing head units.

The innovative design makes maximum use of the space available, providing holding slots for no less than six different sizes of tube and a standard assay plate, while the center pad area can be used for quick hand-held mixing of a vast range of tubes and vessels.
  • Body made from durable ABS to resist most commonly used chemicals, and is easy to clean
  • Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
  • Innovative head can be removed for cleaning purposes
  • US Design Patent No. D787,085, US Utility Patent No. 9895670

Spécifications

Orbit 3.7 mm
Load Bearing Capacity (Metric) 135 g
Speed Range 1000 to 3000 rpm
Electrical Requirements 220/240 V, 50/60 Hz
Voltage 220/240 V
Wattage 44 W
Amperage 0.4 A
Tube Capacity 50 mL, 15 mL, 1.5/2 mL, 0.5 mL, and 0.2 mL
Product Type Vortexing unit
Load Bearing Capacity (English) 4.8 oz.
Plug Type CE
Frequency 50/60 Hz
Material ABS Base, Nylon Platform
Depth (English) 7.41 in.
Depth (Metric) 19 cm
Dimensions (L x W) 6.7 x 7.5 in.
Height (English) 7.68 in.
Height (Metric) 19.7 cm
Length (English) 6.7 in.
Length (English) Platform 5.51 in.
Length (Metric) 17.1 cm
Length (Metric) Platform 140 mm
Width (English) 6.67 in.
Width (English) Platform 4.5 in.
Width (Metric) 17.1 cm
Width (Metric) Platform 11.43 cm
Shipping Weight (English) 11.12 lb.
Shipping Weight (Metric) 5 kg
Weight (English) 8.66 lb.
Weight (Metric) 3.93 kg
Color Blue
Description Vortexer Mixer 220/240 V, CE Plug
Model HS120212
For Use With (Equipment) Tubes, Assay Plates
Includes 1 x AC power cord, 1 x mixing head, 3 x elastic retention cords, 3 x central pads
Motor Type Brushless
Operating Modes 3-Modes: 1000, 2000, and 3000 rpm
Warranty and Service Offering 5 Years
Afficher plus de résultats Afficher moins de résultats