missing translation for 'onlineSavingsMsg'
Learn More

Volatiles Mix for OLM04.1, SPEX CertiPrep™

Catalog No. CLPV43CH
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLPV43CH 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLPV43CH Supplier SPEX CertiPrep Group LLC Supplier No. CLPV43CH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,1,1-Trichloroethane71-55-62000μg/mL
1,1,2,2-Tetrachloroethane79-34-52000μg/mL
1,1,2-Trichloroethane79-00-52000μg/mL
1,1,2-Trichlorotrifluoroethane76-13-12000μg/mL
1,1-Dichloroethane75-34-32000μg/mL
1,1-Dichloroethene75-35-42000μg/mL
1,2,4-Trichlorobenzene120-82-12000μg/mL
1,2-Dibromo-3-chloropropane96-12-82000μg/mL
1,2-Dibromoethane106-93-42000μg/mL
1,2-Dichlorobenzene95-50-12000μg/mL
1,2-Dichloroethane107-06-22000μg/mL
1,2-Dichloropropane78-87-52000μg/mL
1,3-Dichlorobenzene541-73-12000μg/mL
1,4-Dichlorobenzene106-46-72000μg/mL
2-Butanone78-93-32000μg/mL
2-Hexanone591-78-62000μg/mL
4-Methyl-2-pentanone108-10-12000μg/mL
Acetone67-64-12000μg/mL
Benzene71-43-22000μg/mL
Bromodichloromethane75-27-42000μg/mL
Bromoform75-25-22000μg/mL
Carbon Disulfide75-15-02000μg/mL
Carbon tetrachloride56-23-52000μg/mL
Chlorobenzene108-90-72000μg/mL
Chloroform67-66-32000μg/mL
cis-1,2-Dichloroethene156-59-22000μg/mL
cis-1,3-Dichloropropene10061-01-52000μg/mL
Cyclohexane110-82-72000μg/mL
Dibromochloromethane124-48-12000μg/mL
Ethylbenzene100-41-42000μg/mL
Isopropylbenzene98-82-82000μg/mL
Methyl acetate79-20-92000μg/mL
Methylcyclohexane108-87-22000μg/mL
Methylene chloride75-09-22000μg/mL
m-Xylene108-38-32000μg/mL
o-Xylene95-47-62000μg/mL
p-Xylene106-42-32000μg/mL
Styrene100-42-52000μg/mL
tert-Butyl methyl ether1634-04-42000μg/mL
Tetrachloroethene127-18-42000μg/mL
Toluene108-88-32000μg/mL
trans-1,2-Dichloroethene156-60-52000μg/mL
trans-1,3-Dichloropropene10061-02-62000μg/mL
Trichloroethene79-01-62000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Volatiles Mix for OLM04.1
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA CLP Methods
Technique GC-MS
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol-P and T
Quantity 1 mL

Product intended for laboratory use only