missing translation for 'onlineSavingsMsg'
Learn More

Volatile 1 -WS, MilliporeSigma™ Supelco™

Catalog No. 111017026
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017026 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017026 Supplier Merck Emd Millipore Supplier No. PE13131.2ML
Only null left

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C - closed cup
Purity Grade Notes The PT sample is provided as a 1.2 mL concentrate.The sample is methanol.The PT sample is not preserved.The sample must be stored (at 4°C. Only 80% of analytes will be spiked in this sample.)
Type Proficiency Testing Material
Quantity 1.2 mL