missing translation for 'onlineSavingsMsg'
Learn More

VOCs - Clay 4, MilliporeSigma™ Supelco™

Numéro de catalogue. 111001643
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111001643 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111001643 Fournisseur Merck Emd Millipore Code fournisseur CRM63825G
Il en reste null

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Spécifications

CAS 14808-60-7
Type Certified Reference Material
Quantity 25 g
Grade Certified Reference Material