missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Vivaspin™ 500 Sample Concentrators

Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Vivaspin™ 500 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.

$224.98 - $224.98

Specifications

Material (Membrane) PES
Filtration Area 0.5 cm2
Quantity 25/Pk.
Volume (Metric) 500 μL
Length (Metric) 50 mm
View More Specs

Products 6
Catalog Number Mfr. No. MWCO Price Quantity  
Catalog Number Mfr. No. MWCO Price Quantity  
45-001-562
View Documents
Cytiva
28932235
30 kDa
Pack of 25 Each for $224.98
Only null left
 
45-001-559
View Documents
Cytiva
28932218
3 kDa
Pack of 25 Each for $224.98
Only null left
 
45-001-563
View Documents
Cytiva
28932236
50 kDa
Pack of 25 Each for $224.98
Only null left
 
45-001-564
View Documents
Cytiva
28932237
100 kDa
Pack of 25 Each for $224.98
Only null left
 
45-001-561
View Documents Promotion Details
Cytiva
28932225
10 kDa
Pack of 25 Each for $224.98
Only null left
 
45-001-560
View Documents
Cytiva
28932223
5 kDa
Pack of 25 Each for $224.98
Only null left
 
Description

Description

Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.

  • Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
  • Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
  • Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
  • Compatible pH range: 1 to 9
  • Active membrane area: 0.5cm2

For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.

Specifications

Specifications

PES
25/Pk.
50 mm
0.5 cm2
500 μL
11 mm
Videos
SDS
Documents

Documents