missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivapore 10-20 Expansion Reservoir

Catalog No. 01288108
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01288108 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288108 Supplier Sartorius Supplier No. VPARV0010D

Vivapore 10-20 expansion reservoir with simple push-fit handling is compatible with Vivapore 10 solvent absorption concentrators. Can either be used to increase the sample capacity of Vivapore from 10 to 20 mL, or for continuous diafiltration for sample desalting and buffer exchange applications.

  • Concentrate Equipment-Free: Solvent absorption is ideal where centrifuges are not available and for high-throughput applications.
  • Fill and Go: Simply add your sample and leave it to reach the desired final volume, with no risk of concentration to dryness.
  • Tailored Process Monitoring: Printed markings help to reduce the guesswork when each sample needs to reach its own specific concentration factor.
  • Maintained Sample Integrity: Self-contained, single use units ensure contamination-free concentration of individual samples.
  • Sensitive Molecule Friendly: Benchtop or refrigerated low pressure processing for shear and temperature sensitive molecules.
  • Expand or Exchange: Optional reservoirs provide the flexibility to double sample capacity or concentrate and desalt in one step

Specifications

Material Polypropylene
Sterility Non-sterile
Type Reservoir
Volume (Metric) 10 mL
Sample Volume 10 mL