missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K1, MP Biomedicals™

Catalog No. ICN10328301
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10328301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10328301 Supplier MP Biomedicals Inc Supplier No. 0210328301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84-80-0
Molecular Formula C31H46O2
Molecular Weight (g/mol) 450.707
InChI Key MBWXNTAXLNYFJB-NKFFZRIASA-N
Synonym vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone
PubChem CID 5284607
ChEBI CHEBI:18067
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
SMILES CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Color Yellow
Density 0.984
Quantity 1 g
Formula Weight 450.68
Physical Form Viscous Liquid
Chemical Name or Material Vitamin K1