missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin A acetate, MP Biomedicals™
$54.10 - $174.13
Chemical Identifiers
| CAS | 127-47-9 |
|---|---|
| Molecular Formula | C22H32O2 |
| Molecular Weight (g/mol) | 328.496 |
| MDL Number | MFCD00019413 |
| InChI Key | QGNJRVVDBSJHIZ-QHLGVNSISA-N |
| Synonym | retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650 |
| PubChem CID | 638034 |
| ChEBI | CHEBI:32095 |
| IUPAC Name | [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate |
| SMILES | CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10325710
|
N/A | 10 g |
Each for $54.10
|
|
|||||
|
ICN10325725
|
N/A | 25 g |
Each for $72.85
|
|
|||||
|
ICN10325780
|
N/A | 100 g |
Each for $174.13
|
|
|||||
Chemical Identifiers
| 127-47-9 | |
| 328.496 | |
| QGNJRVVDBSJHIZ-QHLGVNSISA-N | |
| 638034 | |
| [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate |
| C22H32O2 | |
| MFCD00019413 | |
| retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650 | |
| CHEBI:32095 | |
| CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C |
Specifications
| 127-47-9 | |
| C22H32O2 | |
| retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650 | |
| CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C | |
| 328.496 | |
| CHEBI:32095 | |
| Crystalline Powder |
| Yellow | |
| MFCD00019413 | |
| QGNJRVVDBSJHIZ-QHLGVNSISA-N | |
| [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate | |
| 638034 | |
| 328.5 | |
| Vitamin A acetate |
Safety and Handling
EINECSNumber : 204-844-2
Recommended Storage : Store at +4°C