missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A acetate, MP Biomedicals™

Catalog No. ICN10325710
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10325710 10 g
ICN10325725 25 g
ICN10325780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10325710 Supplier MP Biomedicals Inc Supplier No. 0210325710

Chemical Identifiers

CAS 127-47-9
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.496
MDL Number MFCD00019413
InChI Key QGNJRVVDBSJHIZ-QHLGVNSISA-N
Synonym retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
PubChem CID 638034
ChEBI CHEBI:32095
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C

Specifications

Color Yellow
Quantity 10 g
Formula Weight 328.5
Physical Form Crystalline Powder
Chemical Name or Material Vitamin A acetate
EINECSNumber 204-844-2
Recommended Storage Store at +4°C