missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A acetate, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210325725

Quantity 25 g

 View more versions of this product

Catalog No. ICN10325725


Only null left

Description

Description

Specifications

Chemical Identifiers

127-47-9
328.496
QGNJRVVDBSJHIZ-QHLGVNSISA-N
638034
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
C22H32O2
MFCD00019413
retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
CHEBI:32095
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C

Specifications

Vitamin A acetate
Yellow
25 g
retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C
328.496
CHEBI:32095
Crystalline Powder
127-47-9
C22H32O2
MFCD00019413
QGNJRVVDBSJHIZ-QHLGVNSISA-N
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
638034
328.5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-844-2

Recommended Storage : Store at +4°C

SDS
Documents

Documents

Promotions

Promotions