missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Brand Precision Pipetters
GREENER_CHOICE

Numéro de catalogue. p-49368
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
150 μL
200 μL
500 μL
1000 μL
Color:
Black
Blue
Gray
Green
Orange
Red
Silver
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric) Color
21320 10 μL Orange
21322 20 μL Black
21324 25 μL White
21326 50 μL Green
21328 100 μL Blue
2138162 150 μL Silver
21330 200 μL Red
21336 500 μL Yellow
21338 1000 μL Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Numéro de catalogue. 21320 Fournisseur VistaLab™ Technologies Code fournisseur VIS1021
Il en reste null

Ten fixed-volume models from 10 to 1000μL

  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet

Order Info

Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.

Spécifications

Accuracy ±5%
Autoclavable Not Autoclavable
Color Orange
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum, Stainless Steel
Model F1
Number of Channels 1
Sterility Non-sterile
Type Precision Pipette
Volume (Metric) 10 μL
Includes Nozzle Insert, Calibration Key
Quantity 1 Ea.
Warranty Limited Lifetime
Length (English) 9 in.
Width (English) 2 in.
Height (English) 1.5 in.
Afficher plus de résultats Afficher moins de résultats