Learn More
Leica Microsystems Visoria M Materials Microscope with Flexacam c5 for Inspection and Measurement of Fine Details and Surface Structures
Numéro de catalogue. 14823442
Change view
Click to view available options
Magnification Power:
5X to 100X
Numéro de catalogue. Magnification Power
14823442 5X to 100X
1 options
Numéro de catalogue. 14823442 Fournisseur Leica Microsystems Code fournisseur 111000009
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This microscope is suitable for metal, electronics, and polymer industries, and materials science labs. Ideal for inspecting and measuring fine details and surface structures of samples and workflows can be streamlined with encoded functions, optimized light settings, and other features.
  • Visoria M Materials Microscope with illumination management
  • 10 in. screen/tablet
  • Powered by the Enersight microscope software platform (Desktop)
  • Flexacam i5 microscope camera, including power supply
  • High-power white LED
  • Incident light axis, 4-position
  • Contrast methods (incident light): Brightfield, Darkfield, Oblique illumination
  • Encoded nosepiece, 5x
  • Objectives (N PLAN EPI): 5x, 10x, 20x, 50x, 100x
  • Manual XY-stage (76 x 50 mm)
  • 3-step focus drive with height-adjustable focus knobs
  • Dust protection cover
  • Quick start guide and user manual
  • Efficiency through encoded functions
  • More comfort with ergonomics
  • Work without eyepieces by going digital
  • Visualize sample details with the right contrast
Spécifications
Eyepiece 10X/22 mm
Head Style Trinocular, 30° Inclined
Lighting Type LED
Magnification Power 5X to 100X
Model Visoria M
Objective Types Plan
Product Type Darkfield Microscope