Learn More
Thermo Scientific™ Virtuoso™ Vial Identification System Collection Tray
Catalog No. 03375309
Change view
Click to view available options
For Use With (Equipment):
Virtuoso Identification System
Catalog No. For Use With (Equipment)
03375309 Virtuoso Identification System
1 options
Catalog No. 03375309 Supplier Thermo Scientific™ Supplier No. 60180VT502
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use this tray to collect vials after they are labeled by the Thermo Scientific™ Virtuoso™ Vial Identification System.
Use the Thermo Scientific™ Virtuoso™ Vial Identification System Collection Tray to collect vials in a central location beneath the unit after vial printing. The Virtuoso Vial Identification System automates vial labeling and ensures accurate vial identification and reliable data, and increases laboratory productivity.
Specifications
Product Line Virtuoso
Description Virtuoso Vial Collection Tray
For Use With (Application) Vial ID and Labelling
For Use With (Equipment) Virtuoso Identification System
No. per Pack 3/Pk.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.