missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViralSEQ™ dPCR AAV Titer Kit
GREENER_CHOICE

Catalog No. A59364
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A59364 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A59364 Supplier Applied Biosystems™ Supplier No. A59364
Get an Extra Analytical Consumable with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems ViralSEQ dPCR AAV Titer Kit is a digital PCR (dPCR) assay for the reproducible quantitation of AAV particles independent of the serotype.

The Applied Biosystems ViralSEQ dPCR AAV Titer Kit is a digital PCR (dPCR) assay for the reproducible quantitation of AAV particles independent of the serotype. AAV titer quantitation is a critical quality attribute (CQA) in viral vector manufacturing for gene therapy development. This probe-based dPCR assay delivers superior sensitivity and consistency, outperforming conventional DNA blotting or ELISA methods.

Features of the ViralSEQ dPCR AAV Titer Kit include:

  • Accurate quantitation of viral particles by dPCR targeting ITR and SV40
  • Assay design compatible with more than 80% of AAV transfer plasmids
  • Easy-to-use, integrated sample-to-results system features TaqMan dPCR master mix and TaqMan primer/probe set
  • Highly sensitive quantitation delivers results in about three hours
  • Optional manual or automated sample preparation, optimized for quantitative recovery from bioproduction samples

The high sensitivity and specificity provided by the ViralSEQ dPCR AAV Titer Kit make it suitable for viral quantitation in process development and lot release testing.

Digital PCR workflow
The ViralSEQ dPCR AAV Titer Kit is part of an integrated dPCR workflow for viral vector characterization during biopharmaceutical manufacturing. Optional use of the Pharma KingFisher Apex 96 Deep-Well Magnetic Particle Processor with the PrepSEQ Nucleic Acid Sample Preparation Kit helps ensure high recovery of viral DNA with less labor and error than manual processing. The Pharma KingFisher Apex instrument can process up to 24 samples in triplicate, compared to three samples in triplicate using a manual method.

To help ensure performance that meets or exceeds regulatory compliance, the ViralSEQ kit has been internally validated on the QuantStudio Absolute Q Digital PCR System. Data analysis is streamlined using the QuantStudio Absolute Q dPCR Software, which includes accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Part 11 compliance.

Specifications

Content And Storage • 5X Absolute Q Digital PCR Master Mix, 0.36 mL; Store at -15°C to -25°C, protect from light
• dPCR AAV Titer Assay Mix, 90 μL; Store at -15°C to -25°C, protect from light
• Negative Control (water), 1 mL; Store at -15°C to -25°C before first use and 2°C to 8°C after first use
Detection Method Digital PCR
Form Frozen
PCR Method Digital PCR
Label or Dye FAM, VIC, ABY
Product Line ViralSEQ
Product Type dPCR AAV Titer Kit
Quantity 100 reactions
For Use With (Application) Virus Detection
No. of Reactions 100 Reactions

For Research Use Only.