missing translation for 'onlineSavingsMsg'
Learn More

Vinylmagnesium chloride, 1.5M (13 wt.%) solution in THF, AcroSeal™

Catalog No. AC252591000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC252591000 100 mL AcroSeal™ Glass bottle
AC252598000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC252591000 Supplier Thermo Scientific Chemicals Supplier No. 252591000
Only null left

CAS: 3536-96-7 | C2H3ClMg | 86.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3536-96-7 , 109-99-9
Molecular Formula C2H3ClMg
Molecular Weight (g/mol) 86.80
MDL Number MFCD00013594
InChI Key NAFZXKWEUWVSAW-UHFFFAOYSA-M
Synonym vinylmagnesium chloride, chlorovinylmagnesium, chloro ethenyl magnesium, vinyl magnesium chloride, magnesium, chloroethenyl, vinyl magnesium-chloride, vinyl-magnesium chloride, magnesium,chloroethenyl, ethenyl magnesium chloride, vinylmagnesium chloride solution
PubChem CID 77075
IUPAC Name magnesium;ethene;chloride
SMILES Cl[Mg]C=C

Specifications

Color White to Beige
Boiling Point 66.0°C
Linear Formula CH2=CHMgCl
Density 0.9800g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 86.8
Percent Purity 2M (18 wt.%)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.98
Quantity 100 mL
Flash Point −017°C
Physical Form Crystalline Powder
Chemical Name or Material Vinylmagnesium chloride, 15 wt. % Solution in Tetrahydrofuran
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 222-575-9
Recommended Storage Below 10°C, crystallization of Grignard reagent will occur. Store between 10°C and 25°C.

RUO – Research Use Only