missing translation for 'onlineSavingsMsg'
Learn More

Vinylboronic acid pinacol ester, 97+%, stabilized with 0.05% BHT

Catalog No. AC395530010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395530010 1 g Glass bottle
AC395530050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC395530010 Supplier Thermo Scientific Chemicals Supplier No. 395530010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75927-49-0 | C8H15BO2 | 154.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75927-49-0
Molecular Formula C8H15BO2
Molecular Weight (g/mol) 154.02
MDL Number MFCD00192492
InChI Key DPGSPRJLAZGUBQ-UHFFFAOYSA-N
Synonym vinylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-vinyl-1,3,2-dioxaborolane, vinylboronic acid pinacol cyclic ester, 2-vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-ethenyl-4,4,5,5-tetramethyl, pinacol vinylboronate, stabilized with bht, 2-vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaoborolane, vbpe, pubchem23743, ethenylboronic acid pinacol ester
PubChem CID 5233012
IUPAC Name 2-ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C=C

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color White to Yellow
Density 0.9080g/mL
Boiling Point 40°C (20.0 mmHg)
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.908
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Stabilizer 500ppm (BHT) max.
Formula Weight 154.02
Percent Purity 97+%
Physical Form Powder or Low Melting Solid
Chemical Name or Material Vinylboronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only